ContactUsLinkComponent
Arsenic limit test apparatus
Arsenic limit test apparatus
Catalog # 637-0000
Supplier:  DIXON SCIENTIFIC
undefined
Arsenic limit test apparatus
Catalog # 637-0000
Supplier:  DIXON SCIENTIFIC
Specifications
  • Description:
    Arsenic limit test apparatus
  • Pk:
    1
Specifications
About this item
The two part apparatus is designed to hold a test paper between two flat ground flanges that are held together by two spiral springs. The lower tube part has a 19/26 ground glass joint suitable for fitting a 100 ml conical flask. The gases evolved from the flask diffuse up the tube through the pre-treated filter paper, which captures the arsine and results in a yellow coloured stain.
Certifications: Conforms to British Pharmacopoeia (2003), Ph.Eur. (2005) requirements
Delivery: Supplied without conical flask.