Capillary columns, BPX70
Supplier: Trajan Scientific and Medical
Certificates
About this item
Polarity has been designed for fatty acid methyl ester (FAME) analysis.
70% cyanopropyl polysilphenylene-siloxane.
Max. temperature: 260 °C
Suitable replacement for: CP-Sil88, DB-23, HP-23, Rtx-2330, SP-2330, SP-2380.
Frequently Bought Together