ContactUsLinkComponent
Clamps, three pronged
Clamps, three pronged
Catalog # 212-9500
Supplier:  R L ENTERPRISES
undefined
Clamps, three pronged
Catalog # 212-9500
Supplier:  R L ENTERPRISES
Specifications
  • Spanning width:
    70 mm
  • Description:
    Clamps, three pronged
  • Pk:
    1
Specifications
About this item
Aluminium die-cast clamps with three rubber covered jaws and nickel plated screws.