Bottles, wide mouth, with screw cap, high performance
Supplier: VWR Collection
Certificates
About this item
LDPE, HDPE or PP with PP screw cap.
- Good chemical resistance
- LDPE and HDPE heat resistant up to 80 °C
- PP heat resistant up to 135 °C, autoclavable
- Leakproof
Can be used anywhere in the laboratory, as well as for collecting samples. Wide opening allows rapid filling and dispensing of powders, pastes and liquids. 4 litre bottles have integrated handle design.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Related Information
Certificates
Search by Lot NumberCoC Bottles wide neck with screw cap High Performance.pdf CoC VWR® Bottles Wide Neck with Screw Cap High Performance CoC_VWRI215-1889_VBP170821 CoC_VWRI215-1889_VMP270820 CoC_VWRI215-1889_VXP040520
More
VWR Large Wide mouth Round Bottles VWR PL Bottles Drawing
Technical Drawing
VWR-Narrow-mouth-and-wide-mouth-bottles-215-3400-to-215--3454.pdf
Product specification
VWRI215-3454S_TDS_EN
Datasheet
VWR® Lab bric-a-brac
Product Literature
Frequently Bought Together