Bottles, wide neck, with thread
Supplier: KJS KUNSTSTOFFVERPACKUNGEN
Certificates
About this item
LDPE, natural colour.
- Good chemical resistance
- Leakproof, tough and versatile
Perfect for sample collection, storage and transport. Ideal for liquids, paste-like media and granular material. The wide mouth allows easy filling and cleaning. Without graduation.
Ordering information: Please order screw caps separately.
Screw caps
Product Information
Frequently Bought Together