Order Entry
Northern Ireland
ContactUsLinkComponent
 
Bottles, narrow neck, round, LDPE, with screw caps
  215-2713
undefined
Bottles, narrow neck, round, LDPE, with screw caps
  215-2713

 

  • Ext.Ø:
    70 mm
  • Capacity:
    500 ml
  • Height:
    172 mm
  • Neck I-Ø:
    18 mm
  • Pk:
    10

 

 

LDPE, natural colour.

  • Chemical resistant, black screw cap, leakproof
  • Good chemical resistance, good thermal resistance
  • With specially reinforced shoulder