Methylthymol blue sodium salt indicator for metal titration, Fluka™
Catalog # 31042-5G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Identity (IR):complying
- Appearance of the solution:complying
- Suitability for Complexometry:complying
- Absorption maximum:433 - 443 nm
- Absorbtion (1%/1cm):90 - 130
Specification Test Results
About this item
:
:844.76 g/mol
Storage Temperature:Ambient
EINECS:217-743-3
CAS Number:1945-77-3
Formula:C₃₇H₄₀N₂Na₄O₁₃S
MDL Number:MFCD00148905