


Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v)
Catalog # A0944.0500
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
UN:2810
ADR:6.1,II
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
CAS Number:136112-00-0
MDL Number:MFCD00133763