

Triethyl orthoacetate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-39-7
:
Formula:C₈H₁₈O₃
MDL Number:MFCD00009223
:162.23 g/mol
Boiling Pt:142 °C (760 mmHg)
Density:0,885 g/cm³
Flash Pt:36 °C
Storage Temperature:Ambient
EINECS:201-112-4
UN:3272
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3950 to 1.3980 (20°C, 589 nm)
- GC:≥96.0 %
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together