Order Entry
Northern Ireland
ContactUsLinkComponent
1,2-Diaminocyclohexanetetraacetic acid monohydrate ≥98.5% for complexometry, Fluka™
1,2-Diaminocyclohexanetetraacetic acid monohydrate ≥98.5% for complexometry, Fluka™
Certificates
1,2-Diaminocyclohexanetetraacetic acid monohydrate ≥98.5% for complexometry, Fluka™
Certificates

Specification Test Results

  • Assay (calculated on the anhydrous basis):
    Min. 99 %
  • Water (Karl Fischer):
    4.8 - 5.6 %
  • Iron (Fe):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.01 %

Specification Test Results

Related Information

Frequently Bought Together