Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ZZW-115 trihydrochloride
ZZW-115 trihydrochloride ≥95%
ZZW-115 trihydrochloride ≥95%
ZZW-115 trihydrochloride ≥95%
Catalog # CAYM34974-1
Supplier:
Cayman Chemical
New Product
CAS Number:
Expand image
ZZW-115 trihydrochloride ≥95%
Catalog # CAYM34974-1
Supplier:
Cayman Chemical
New Product
CAS Number:
About this item
Product Details
Product Details