ContactUsLinkComponent
Uvex Planet Safety Spectacle
Uvex Planet Safety Spectacle
Catalog # UVEX9128295
Supplier:  UVEX
New Product
Uvex Planet Safety Spectacle
Catalog # UVEX9128295
Supplier:  UVEX
New Product
Specifications
  • Colour:
    Green/grey
  • Description:
    Planet safety spectacle
  • Lenses:
    Clear
  • Pk:
    1
Specifications
About this item
Attractive, classic, sporty design, pleasantly lightweight and optimal protection - the uvex pheos nxt. The special design of the arms not only looks impressive: the direction-focused structure also provides extremely good grip - even during demanding tasks. Numerous openings in the side arms improve air circulation and make the glasses light yet no less robust. The large panoramic lens also provides a wide field of vision free from distortion to ensure a good view.
  • Classic, sporty and lightweight with innovative arm design for an optimal, reliable fit
  • uvex protecting planet - mass-balanced PC lens, arms and packaging partially made from recycled material
  • Robust polycarbonate lens with proven uvex supravision coating technology - permanently anti-fog and scratch-resistant lens
  • Metal-free
  • Frame colour: Anthracite, jade
  • PC lens: clear
  • UV400 (UV protection)
  • Anti-fog on the inside
  • Extremely scratch-resistant
  • Chemical-resistant on the outside
Ergonomically shaped arms with grip structure and soft ear-pieces ensure a comfortable and secure fit without creating pressure points. Increased air circulation for outstanding, climate-optimised wearer comfort through numerous openings in the side arms. Anatomically shaped nose piece for all-day wearer comfort.
Certifications: Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters). Certification: W 166 FT CE – 2C-1.2 W1 FTKN CE. UV400 – 100% protection against harmful UV radiation to 400 nm.