Potassium permanganate 30 g/l in ortho-phosphoric acid 22% Reag. Ph. Eur. 1070901
Catalog # 87918.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:1805
ADR:8,III
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364