Phenylhydrazinium chloride 0,36% in hydrochloric acid 5% Reag. Ph. Eur. 1064501
Catalog # 87892.230
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 mL
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride
:
:144.6 g/mol
EINECS:200-444-7
UN:1789
ADR:8,III
CAS Number:59-88-1
Formula:C₆H₉ClN₂
MDL Number:MFCD00012924