Order Entry
Northern Ireland
ContactUsLinkComponent
 
Flammable
Corrosive
Irritant
Ninhydrin solution R3 Reag. Ph. Eur. 1058306
Catalog # 87879.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Ninhydrin solution R3 Reag. Ph. Eur. 1058306
Catalog # 87879.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :178.14 g/mol
Storage Temperature:Ambient
EINECS:207-618-1
UN:1993
ADR:3,III
 :GHS02
 :GHS05
 :GHS07
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791