

Gadolinium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:94219-55-3
:
Formula:Gd(NO₃)₃·xH₂O
MDL Number:MFCD00149730
Merck Index:14,4326
Melting Pt:92 °C
Density:2,406
EINECS:233-437-2
UN:1477
ADR:5.1,III
:343.26 g/mol
:GHS03
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together