



Ammonia 2 M in methanol, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7664-41-7
:
Formula:NH₃
MDL Number:MFCD00011418
:17.03 g/mol
Density:0,787 g/cm³
Flash Pt:6 °C
Storage Temperature:Refrigerator
EINECS:215-647-6
UN:1992
ADR:3,II
:GHS06
:GHS08
:GHS02
:GHS05
Specification Test Results
- Appearance:Clear colorless Solution
- Water:≤0.3 %
- Strength:1.90 to 2.20 M
Specification Test Results
Related Information
Frequently Bought Together