Aluminium lanthanum (III) oxide substrate 10x10x0.5 mm, polished both sides 100 orientation
Catalog # 41434.KT
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
CAS Number:12003-65-5
:
Formula:AlLaO₃
MDL Number:MFCD00144550
:213.89 g/mol
Storage Temperature:Ambient
EINECS:234-433-3