Order Entry
Northern Ireland
ContactUsLinkComponent
1-Chloro-3-methoxy-2-propanol, Sigma-Aldrich®
1-Chloro-3-methoxy-2-propanol, Sigma-Aldrich®
Catalog # 8.14513.0025
Supplier:  Merck
CAS Number:  
1-Chloro-3-methoxy-2-propanol, Sigma-Aldrich®
Catalog # 8.14513.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    124.56 g/mol
  • Boiling Point:
    171 °C
  • Flash Point:
    72 °C
  • Density:
    1.16 g/cm3 (20 °C)
  • pH:
    5.6 (50 g/l, H2O, 20 °C)

Specification Test Results

About this item

Synonyms: 1-Chloro-3-methoxypropan-2-ol, 3-Chloro-1-methoxy-2-propanol
 :
 :124.57 g/mol
Boiling Pt:171 °C (1013 hPa)
Density:1,16 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:223-982-4
 :GHS07
Storage Temperature:Ambient
CAS Number:4151-97-7
Formula:C₄H₉ClO₂
MDL Number:MFCD00018935