Standard for ViscoQC Viscometers
PAAR191118
New Product
- Description:Silicone viscosity standard AP RT5k, 5,6/5,0 Pa.s at 20/25 °C
- Environmentally Preferable:
- Pk:600 mL
Silicone viscosity standard for use with ViscoQC.
Compatible with: ViscoQC 100 H, ViscoQC 100 L, ViscoQC 100 R, ViscoQC 300 H, ViscoQC 300 L, ViscoQC 300 R