Laboratory Washer-Disinfector, PG 8582
Catalog # MIEL9960770
Supplier: MIELE
New Product
Specifications
- Circulation pump (Qmax):500 L/min
- Description:Washer-disinfector, PG 8582 WW AD LD, with liquid dispensing and EcoDry function.
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Washer-disinfector with liquid dispensing and EcoDry function.
- Performance/cycle, e.g. 8 DIN mesh trays or 42 GYN specula
- Safety through monitoring - wash pressure and spray arm monitoring
- Convenient drying support - EcoDry
- Total security - link to process documentation possible