Anti-IGHG1 Rabbit Polyclonal Antibody (PerCP)
NOVUNBP2-24891PCP
New Product
- Antigen symbol:IGHG1
- Clonality:Polyclonal
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit
- Antigen synonyms:Immunoglobulin G|ImmunoglobulinG
- Purity:Affinity purified
- Storage buffer:PBS, 0,05% Sodium Azide
- Storage temperature:Store at 4 °C
- Pk:0,1 mL
Type:
Antigen: IGHG1
Clonality: Polyclonal
Clone:
Conjugation: PerCP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit