Order Entry
Northern Ireland
ContactUsLinkComponent
1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one 98%
Irritant
1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one 98%
Catalog # 186310010.
CAS Number:  
Irritant
1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one 98%
Catalog # 186310010.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    white to beige powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :162.19 g/mol
EINECS:233-148-1
 :GHS07
Storage Temperature:Ambient
CAS Number:10045-45-1
Formula:C₉H₁₀N₂O
MDL Number:MFCD00005715