Order Entry
Northern Ireland
ContactUsLinkComponent
Single Valve SPE Cartridge
Single Valve SPE Cartridge
Catalog # PERCN2991112
Supplier:  PerkinElmer
New Product
Single Valve SPE Cartridge
Catalog # PERCN2991112
Supplier:  PerkinElmer
New Product

Specifications

  • Ø int.:
    2 mm
  • Description:
    Single valve SPE cartridge
  • Length:
    10 mm
  • Pk:
    1

Specifications

About this item

The online SPE cartridge has been specially designed for elution to the HPLC column. Dimensions are optimised to combine high extraction capacity with small elution volumes.

Dimension 10×1 mm is also available.