

Mycophenolic acid-O-β-D-glucuronide solution Acetonitrile:Water (90:10) (v/v) 1 mg/ml, Cerilliant®, certified reference material
Catalog # M-250-1ML
Supplier: Merck
New Product
CAS Number:
Specifications
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:496.461 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ300
Flash Pt:2,0 °C (closed cup)
UN:1648
:GHS02
:GHS07
Storage Temperature:Freezer
CAS Number:31528-44-6
Formula:C₂₃H₂₈O₁₂
MDL Number:MFCD09878308