C-4 ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 517301-10
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
:407.85 g/mol
CAS Number:864426-22-2
Formula:C₂₃H₁₈ClNO₄