
Sodium metaarsenite 0.05 mol/l (0.1 N), AVS TITRINORM, CRM (Certified Reference Material acc. ISO 17034) traceable to NIST Standard Reference Material (SRM)
Catalog # 92513.290
Supplier: VWR Chemicals
New Product
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
Specifications
About this item
:
:129.91 g/mol
Storage Temperature:Ambient
EINECS:232-070-5
UN:1686
ADR:6.1,II
:GHS08
CAS Number:7784-46-5
Formula:NaAsO₂
MDL Number:MFCD00003472