About this item
CleanDox® is a poly-infused bond paper that can be used throughout your facility for note taking, printing batch records, forms and logbooks.
- Non-sterile and ISO 5 Cleanroom processed
- Vacuum packed with double layers cleanroom bag, packed in controlled ISO 5 cleanroom environment
- Low in chloride and other ionic contamination
- Formulated without magnesium silicate
- Excellent IPA resistant material
- Compatible with most inks used for writing without smearing
- Ultra low ionic contamination, ultra low particle and fiber generation
- Excellent heat resistant material for laser printing and photocopies
Available in 21,59×27,94 cm and A4 sizes. Available in white, blue, green, cream, and pink. Can be autoclaved.
This IPA resistant material is available in several different colours.