Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330 Enol-Glucoside
Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330 Enol-Glucoside
Catalog # 32487-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330 Enol-Glucoside
Catalog # 32487-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    1172614-86-6
  • Description:
    Spirotetramat Metabolite BYI08330 enol-glucoside
  • Seq:
    01
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H411
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    cis-3-(2,5-Dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro [4.5]dec-3-en-4-yl β-ᴅ-glucopyranoside
  • Formula:
    C₂₄H₃₃NO₈
  • Molecular weight:
    463,52
  • Storage temperature:
    −20 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS09

Spirotetramat may be used as a reference standard for QuEChERS based methodology for the determination of spirotetramat and its relevant metabolites in pistachio samples by liquid chromatography-tandem mass spectrometry.

Caution: P273