Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330-Mono-Hydroxy
Catalog # 32486-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1172134-12-1
- Description:Spirotetramat Metabolite BYI08330-mono-hydroxy
- Seq:04
- Storage conditions:–20 °C
- Hazard class description:H411
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Synonym(s):cis-3-(2,5-Dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5]decan-2-one
- Formula:C₁₈H₂₅NO₃
- Molecular weight:303,40
- Storage temperature:−20 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS09
Spirotetramat may be used as a reference standard for QuEChERS based methodology for the determination of spirotetramat and its relevant metabolites in pistachio samples by liquid chromatography-tandem mass spectrometry.
Caution: P273 - P391 - P501