Supelco® Organic Reference Standard, Magnesium Perchlorate Hydrate
Catalog # 31419-100MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard, for GPC, Mw 25000
- Description:Magnesium perchlorate hydrate
- Seq:13
- Storage conditions:2...30 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:Not applicable