Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Dextran
  00893-100MG
 :  Merck
 :  
Supelco® Organic Reference Standard, Dextran
  00893-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard, for GPC, 150000
  • CAS No.:
    9004-54-0
  • Description:
    Dextran
  • Seq:
    17
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00130935
  • M-Clarity quality level:
    MQ100
  • Formula:
    [C₆H₁₀O₅]ₙ