Supelco® PESTANAL® Organic Reference Standard, Diphenyl Sulfone
Catalog # 45458-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:127-63-9
- Description:Diphenyl sulfone
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00007548
- M-Clarity quality level:MQ100
- Synonym(s):Phenyl sulfone
- Melting point:123...129 °C (lit.)
- Formula:(C₆H₅)₂SO₂
- Molecular weight:218,27
- Flash point:184 °C
- Boiling point:379 °C (lit.)
Specifications
About this item
- GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501