Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, 1-(2-Chlorophenyl)-1-(4-Chlorophenyl)-2,2-Dichloroethane
  44012-50MG
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, 1-(2-Chlorophenyl)-1-(4-Chlorophenyl)-2,2-Dichloroethane
  44012-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • CAS No.:
    53-19-0
  • Description:
    1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H351
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    (2,4′-Dichlorodiphenyl)dichloroethane; o,p′-DDD; Mitotane
  • Formula:
    (ClC₆H₄)₂CHCHCl₂
  • Molecular weight:
    320,04

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
 : P280