Supelco® Organic Reference Standard, 2-Chloro-1,3-Propanediol
Catalog # 16761-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:497-04-1
- Description:2-Chloro-1,3-propanediol
- Storage conditions:2...8 °C
- Hazard class description:H301
- Shipping conditions:STD
- MDL No.:MFCD01698127
- M-Clarity quality level:MQ100
- Synonym(s):2-MCPD; 2-Monochloro-propanediol; Glycerol β-chlorohydrin
- Molecular weight:110,54
Specifications
About this item
Caution: P264 - P270 - P301 + P310 - P405 - P501