Supelco® Organic Reference Standard, CARB Method 1004 DNPH Mix 2
Catalog # CRM47651
Supplier: Merck
CAS Number:
Specifications
- Concentration:30 µg/ml in acetonitrile (as the aldehydes and ketones)
- Pk:1 mL
- Standards:Certified reference material
- Description:CARB Method 1004 DNPH mix 2
- Seq:2
- Storage conditions:2…8 °C
- Hazard class description:H225 - H302 + H312 + H332 - H319
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Flash point:2,0 °C - Closed cup
- Pk Info:Ampule of 1×1 ml
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338