Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Cerilliant® Organic Reference Standard, Lofexidine-D₄ hydrochloride solution
Supelco® Cerilliant® Organic Reference Standard, Lofexidine-D₄ hydrochloride solution
Catalog # L-056-1ML
Supplier:  Merck
CAS Number:  
Supelco® Cerilliant® Organic Reference Standard, Lofexidine-D₄ hydrochloride solution
Catalog # L-056-1ML
Supplier:  Merck
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • CAS No.:
    1206845-57-9
  • Description:
    Lofexidine-D₄ hydrochloride solution
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD28899117
  • Molecular weight:
    299,62
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338