Supelco® TraceCERT® Organic Reference Standard, Triazophos
Catalog # CRM74120-50MG
Supplier: Merck
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:24017-47-8
- Description:Triazophos
- Storage conditions:–20 °C
- Hazard class description:H301 - H312 - H330 - H410
- Shipping conditions:WICE
- MDL No.:MFCD00055327
- M-Clarity quality level:MQ300
- Molecular weight:313,31
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P260 - P264 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310