Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, BDE No 209 solution
Supelco® Organic Reference Standard, BDE No 209 solution
Catalog # 89407-1ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, BDE No 209 solution
Catalog # 89407-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    ~50 µg/ml (Isooctane:Toluene 50:50)
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1163-19-5
  • Description:
    BDE No 209 solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H304 - H315 - H336 - H361d - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00000059
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Decabromodiphenyl ether solution; Decabromodiphenyl oxide
  • Molecular weight:
    959,17

Specifications

About this item

The customer needs to make sure that the standard is homogeneous this can be achieved by shaking vortexing or sonicating the ampoule before opening the ampoule.

Care should be taken when opening the ampoule to avoid any injuries from the broken glass. The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent. After opening the ampoule the standard should not be stored or kept in the ampoule.

To preserve the integrity of the product the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.

Caution: P202 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331