Supelco® Organic Reference Standard, 1,3-Dinitrobenzene
45966-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:99-65-0
- Description:1,3-Dinitrobenzene
- Storage conditions:2...30 °C
- Hazard class description:H300 + H310 + H330 - H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00007222
- M-Clarity quality level:MQ100
- Synonym(s):1,3-DNB
- Melting point:84 - 86 °C (lit.)
- Formula:C₆H₄(NO₂)₂
- Molecular weight:168,11
- Density:1,368 g/ml at 25 °C (lit.)
- Flash point:150 °C - Closed cup
- Boiling point:297 °C (lit.)
- GHS signal: GHS06, GHS08, GHS09
: P262 - P264 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310