XFD514 tyramide
Product Details & Documents
XFD 514 tyramide contains the Alexa Fluor® 514 fluorophore that can be readily detected with the standard rhodamine 6G filter set (Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Tyramide signal amplification (TSA) has proven to be a particularly versatile and powerful enzyme amplification technique with improved assay sensitivity.
Caution: For Research Use Only
- Molecular Weight:938.17 g/mol
Specifications
Specifications
- Pk:200