Order Entry
Northern Ireland
ContactUsLinkComponent
Violet 660 sodium salt, CytoCalcein™ excited at 405 nm
Violet 660 sodium salt, CytoCalcein™ excited at 405 nm
Catalog # AATB21903
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Violet 660 sodium salt, CytoCalcein™ excited at 405 nm
Catalog # AATB21903
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :662.63 g/mol