Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

AATOM™ 594 NHS ester

1 mg
Catalog # AATB2860
Irritant
Supplier:   AAT Bioquest
AATOM™ 594 NHS ester
Catalog # AATB2860
Supplier:  AAT Bioquest
CAS Number:  
AATOM™ 594 NHS ester

1 mg

CAS Number:  
Price
£244.00
Log in for availability
1 * 1 mg
Product Details & Documents

AATOM™ 594 is a bright, red fluorescent dye characterized by a strong absorption, high fluorescence quantum yield, and exceptional thermal and photostability.

  • Stable for 12 months

Quality tested in-house

 : RT

 : Minimize light exposure

 : For Research Use Only

  • MDL Number:MFCD07370576
  • Molecular Weight:1004.22 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.