Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-RFX2 Rabbit Polyclonal Antibody
Anti-RFX2 Rabbit Polyclonal Antibody
Catalog # ANTIA101255-100
Supplier:  Antibodies.com
New Product
undefined
Anti-RFX2 Rabbit Polyclonal Antibody
Catalog # ANTIA101255-100
Supplier:  Antibodies.com
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DNA-binding protein RFX2
  • Antigen symbol:
    RFX2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P48378
  • Antigen synonyms:
    HLA class II regulatory factor RFX2|RFX2_HUMAN|Regulatory factor X 2|trans acting regulatory factor 2|regulatory factor X2|regulatory factor X 2 (influences HLA class II expression)
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    80 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human RFX2 (amino acids 661 - 710)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to RFX2 for IHC and ELISA with samples derived from human.

  • Validated Applications: IHC, ELISA
  • Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen: RFX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human