Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-CD10 Mouse Monoclonal Antibody [Clone: MEM-78] (Biotinylated)
Catalog # ANTIA86628-100
Supplier:  ANTIBODIES.COM
New Product
Anti-CD10 Mouse Monoclonal Antibody [Clone: MEM-78] (Biotinylated)
Catalog # ANTIA86628-100
Supplier:  ANTIBODIES.COM
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Atriopeptidase
  • Antigen symbol:
    CD10
  • Clonality:
    Monoclonal
  • Clone:
    MEM-78
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08473
  • Antigen synonyms:
    Enkephalinase|CD10|Common acute lymphocytic leukemia antigen|DKFZp686O16152|CALLA|Membrane metallo endopeptidase|EC 3.4.24.11|CD10 antigen|EPN
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
  • Storage temperature:
    Store at 2...8 °C. Do not freeze!
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    NALM-6 human pre-B cell line
  • Purification:
    This antibody is conjugated with Biotin-LC-NHS under optimum conditions. The reagent is free of unconjugated Biotin.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [MEM-78] antibody to CD10 (Biotin) for Flow Cytometry and IP with samples derived from Human.

  • Validated applications: Flow cytometry, IP
  • Recommended dilutions: Flow cytometry: 1 µg/ml

Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: MEM-78
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human