Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CD35 Mouse Monoclonal Antibody [Clone: E11] (FITC (Fluorescein Isothiocyanate))
Anti-CD35 Mouse Monoclonal Antibody [Clone: E11] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86042-100
Supplier:  Antibodies.com
New Product
Anti-CD35 Mouse Monoclonal Antibody [Clone: E11] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86042-100
Supplier:  Antibodies.com
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C3 binding protein
  • Antigen symbol:
    CD35
  • Clonality:
    Monoclonal
  • Clone:
    E11
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17927
  • Antigen synonyms:
    CD35|complement component (3b/4b) receptor 1 including Knops blood group system|C3BR|CD 35|complement component (3b/4b) receptor 1 (Knops blood group)|Complement component receptor 1|CD35 antigen|C3b/C4b receptor|C4BR
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Acute monocytic leukemia cells and normal blood monocytes
  • Purification:
    This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [E11] antibody to CD35 (FITC) for Flow Cytometry with samples derived from Human and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD35
Clonality: Monoclonal
Clone: E11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates