Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:ALK
- Antigen symbol:ALK
- Clonality:Recombinant
- Clone:ALK1/7008R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q9UM73
- Antigen synonyms:ALK/EML4 fusion gene, included|anaplastic lymphoma kinase (Ki-1)|CD 246|ALK/NPM1 fusion gene, included|Anaplastic lymphoma kinase|anaplastic lymphoma receptor tyrosine kinase|ALK tyrosine kinase receptor|ALK_HUMAN|Anaplastic lymphoma kinase Ki1
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:80 kDa (hybrid)/200 kDa (wild type)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment corresponding to the cytoplasmic domain of human ALK.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [ALK1/7008R] antibody to ALK for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: ALK
Clonality: Recombinant
Clone: ALK1/7008R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human