Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-PKC iota Mouse Monoclonal Antibody [clone: PRKCI/4912]
Anti-PKC iota Mouse Monoclonal Antibody [clone: PRKCI/4912]
Catalog # ANTIA278352-100
Supplier:  Antibodies.com
New Product
undefined
Anti-PKC iota Mouse Monoclonal Antibody [clone: PRKCI/4912]
Catalog # ANTIA278352-100
Supplier:  Antibodies.com
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    aPKC lambda/iota
  • Antigen symbol:
    PKC iota
  • Clonality:
    Monoclonal
  • Clone:
    PRKCI/4912
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P41743
  • Antigen synonyms:
    KPCI_HUMAN|MGC26534|nPKC iota|nPKCiota|Atypical protein kinase C-lambda/iota|aPKC-lambda/iota|nPKC-iota|DXS1179E|Atypical protein kinase C lambda/iota
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    68 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 100-300, of human PKC iota. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [PRKCI/4912] antibody to PKC iota for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: PKC iota
Clonality: Monoclonal
Clone: PRKCI/4912
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human