Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:AN
- Antigen symbol:PAX6
- Clonality:Recombinant
- Clone:PAX6/7078R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P26367
- Antigen synonyms:FVH1|KIAA0552|D11S812E|AN2|LZTS3|Aniridia type II protein|Leucine zipper putative tumor suppressor 3|MGC17209|AN 2
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:47 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment corresponding to the N terminus, around amino acids 1-300, of human PAX6 protein.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [PAX6/7078R] antibody to PAX6 for Flow Cytometry, IF and WB with samples derived from human.
- Validated Applications: Flow Cytometry, IF, WB
- Recommended Dilutions: Flow Cytometry: 0,5 to 1 µg/million cells, IF: 0,5 to 1 µg/ml, WB: 1 to 2 µg/ml
Type: Primary
Antigen: PAX6
Clonality: Recombinant
Clone: PAX6/7078R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human