Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-CD30 Mouse Recombinant Antibody [Clone: rKi-1/6913]
  ANTIA277936-100
New Product
undefined
Anti-CD30 Mouse Recombinant Antibody [Clone: rKi-1/6913]
  ANTIA277936-100
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    CD 30
  • Antigen symbol:
    CD30
  • Clonality:
    Recombinant
  • Clone:
    rKi-1/6913
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Epitope:
    The epitope for this antibody is within the cytoplasmic region, specifically amino acids 495-595.
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28908
  • Antigen synonyms:
    CD30|KI1|Cytokine receptor CD30|KI 1|Ki-1 antigen|D1S166E|KI 1 antigen|CD30L receptor|CD30 antigen
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    105 - 120 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment corresponding to human CD30 protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant mouse monoclonal [rKi-1/6913] antibody to CD30 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD30
Clonality: Recombinant
Clone: rKi-1/6913
Conjugation: Unconjugated
Epitope: The epitope for this antibody is within the cytoplasmic region, specifically amino acids 495-595.
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human